2,6-Diaminopyridine, 98%
2,6-Diaminopyridine, 98%
2,6-Diaminopyridine, 98%
2,6-Diaminopyridine, 98%
Thermo Scientific Chemicals
2,6-Diaminopyridine, 98%
CAS: 141-86-6 | C5H7N3 | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA112381000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Diaminopyridine
Name Note98%
CAS141-86-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger