1,5-Diaminopentane, 98%
1,5-Diaminopentane, 98%
1,5-Diaminopentane, 98%
1,5-Diaminopentane, 98%
Thermo Scientific Chemicals
1,5-Diaminopentane, 98%
CAS: 462-94-2 | C5H14N2 | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA112320050
Price (MYR)
591.00
BOT
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
591.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 5-Diaminopentane
Name Note98%
CAS462-94-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger