1,8-Diaminooctane, 98%
1,8-Diaminooctane, 98%
1,8-Diaminooctane, 98%
1,8-Diaminooctane, 98%
Thermo Scientific Chemicals
1,8-Diaminooctane, 98%
CAS: 373-44-4 | C8H22Cl2N2 | 217.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA112310250
Price (MYR)
312.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
312.00
EA
Add to cart
Specifications
Chemical Name or Material1, 8-Diaminooctane
CAS373-44-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0