1,10-Diaminodecane, 97%
1,10-Diaminodecane, 97%
1,10-Diaminodecane, 97%
1,10-Diaminodecane, 97%
Thermo Scientific Chemicals
1,10-Diaminodecane, 97%
CAS: 646-25-3 | C10H24N2 | 172.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA112130050
Price (MYR)
306.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
306.00
EA
Add to cart
Specifications
Chemical Name or Material1, 10-Diaminodecane
Name Note97%
CAS646-25-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger