3,5-Diaminobenzoic acid, 98%
3,5-Diaminobenzoic acid, 98%
3,5-Diaminobenzoic acid, 98%
3,5-Diaminobenzoic acid, 98%
Thermo Scientific Chemicals
3,5-Diaminobenzoic acid, 98%
CAS: 535-87-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA112111000
Price (MYR)
614.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
614.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 5-Diaminobenzoic acid
Name Note98%
CAS535-87-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning