3,4-Diaminobenzoic acid, 97%
3,4-Diaminobenzoic acid, 97%
3,4-Diaminobenzoic acid, 97%
3,4-Diaminobenzoic acid, 97%
Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%
CAS: 619-05-6 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA112101000
Price (MYR)
936.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
936.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4-Diaminobenzoic acid
CAS619-05-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0