3,3'-Diaminobenzidine, 98+%
3,3'-Diaminobenzidine, 98+%
3,3'-Diaminobenzidine, 98+%
3,3'-Diaminobenzidine, 98+%
Thermo Scientific Chemicals
3,3'-Diaminobenzidine, 98+%
CAS: 91-95-2 | C12H14N4 | 214.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA112080250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 3'-Diaminobenzidine
Name Note99%
CAS91-95-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger