Cytidine, 99+%
Cytidine, 99+%
Cytidine, 99+%
Thermo Scientific Chemicals
Cytidine, 99+%
CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
Catalog number FSA111812500
Price (MYR)
4,105.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
4,105.00
EA
Add to cart
Specifications
Chemical Name or MaterialCytidine
Name Note99.5%
CAS65-46-3
Absorbance0.43 to 0.47 (A250/A260), 2.05 to 2.15 (A280/A260)
Health Hazard 1HYGROSCOPIC