Cyclopentylamine, 99+%
Cyclopentylamine, 99+%
Cyclopentylamine, 99+%
Cyclopentylamine, 99+%
Thermo Scientific Chemicals
Cyclopentylamine, 99+%
CAS: 1003-03-8 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA111590250
Price (MYR)
306.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
306.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCyclopentylamine
Name Note99+%
CAS1003-03-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.