2-Cyclopentenone, 98%
2-Cyclopentenone, 98%
2-Cyclopentenone, 98%
2-Cyclopentenone, 98%
Thermo Scientific Chemicals
2-Cyclopentenone, 98%
CAS: 930-30-3 | C5H6O | 82.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA111570250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Cyclopentenone
CAS930-30-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0