Cyclohexylbenzene, 98%
Cyclohexylbenzene, 98%
Cyclohexylbenzene, 98%
Cyclohexylbenzene, 98%
Thermo Scientific Chemicals
Cyclohexylbenzene, 98%
CAS: 827-52-1 | C12H16 | 160.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA111295000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclohexylbenzene
Name Note98%
CAS827-52-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.