Cyclohexylamine, 99%
Cyclohexylamine, 99%
Cyclohexylamine, 99%
Cyclohexylamine, 99%
Thermo Scientific Chemicals
Cyclohexylamine, 99%
CAS: 108-91-8 | C6H13N | 99.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number FSA111282500
Price (MYR)
228.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
228.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCyclohexylamine
CAS108-91-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Suspected of damaging fertility.
Toxic in contact with skin.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr