2-Cyclohexen-1-ol, 95%
2-Cyclohexen-1-ol, 95%
2-Cyclohexen-1-ol, 95%
2-Cyclohexen-1-ol, 95%
Thermo Scientific Chemicals
2-Cyclohexen-1-ol, 95%
CAS: 822-67-3 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA111230100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Cyclohexen-1-ol
CAS822-67-3
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0