Cyclohexanone oxime, 97%
Cyclohexanone oxime, 97%
Cyclohexanone oxime, 97%
Cyclohexanone oxime, 97%
Thermo Scientific Chemicals
Cyclohexanone oxime, 97%
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA111205000
Price (MYR)
348.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
348.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCyclohexanone oxime
Melting Point88°C to 92°C
Name Note97%
CAS100-64-1
ColorBeige-Brown to White