Cyclohexanone, 99.8%, extra pure
Cyclohexanone, 99.8%, extra pure
Cyclohexanone, 99.8%, extra pure
Cyclohexanone, 99.8%, extra pure
Thermo Scientific Chemicals
Cyclohexanone, 99.8%, extra pure
CAS: 108-94-1 | C6H10O | 98.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA111190025
Price (MYR)
656.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
656.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclohexanone
Melting Point-47.0°C
CAS108-94-1
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Flammable liquid and vapor.