4-Cyanobenzaldehyde, 98%
4-Cyanobenzaldehyde, 98%
4-Cyanobenzaldehyde, 98%
4-Cyanobenzaldehyde, 98%
Thermo Scientific Chemicals
4-Cyanobenzaldehyde, 98%
CAS: 105-07-7 | C8H5NO | 131.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA110721000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Cyanobenzaldehyde
Name Note98%
CAS105-07-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning