Cumene, 98%, pure
Cumene, 98%, pure
Cumene, 98%, pure
Thermo Scientific Chemicals
Cumene, 98%, pure
CAS: 98-82-8 | C9H12 | 120.195 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA110630025
Price (MYR)
605.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
605.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCumene
Name Note98%
CAS98-82-8
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Harmful if swallowed.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.