m-Cresol, 99%
m-Cresol, 99%
m-Cresol, 99%
Thermo Scientific Chemicals
m-Cresol, 99%
CAS: 108-39-4 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA110585000
Price (MYR)
529.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
529.00
BOT
Add to cart
Specifications
Chemical Name or Materialm-Cresol
CAS108-39-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit