o-Cresol, 99%
o-Cresol, 99%
o-Cresol, 99%
Thermo Scientific Chemicals
o-Cresol, 99%
CAS: 95-48-7 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA110555000
Price (MYR)
586.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
586.00
BOT
Add to cart
Specifications
Chemical Name or Materialo-Cresol
CAS95-48-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0