Coronene, 95%
Coronene, 95%
Coronene, 95%
Thermo Scientific Chemicals
Coronene, 95%
CAS: 191-07-1 | C24H12 | 300.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA110511000
Price (MYR)
386.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
386.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCoronene
Name Note95%
CAS191-07-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: