L(+)-Citrulline, 99%
L(+)-Citrulline, 99%
L(+)-Citrulline, 99%
L(+)-Citrulline, 99%
Thermo Scientific Chemicals
L(+)-Citrulline, 99%
CAS: 372-75-8 | C6H13N3O3 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA110470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL(+)-Citrulline
CAS372-75-8
Melting Point214°C
ColorWhite
EINECS Number206-759-6