Citraconic acid, 99+%
Citraconic acid, 99+%
Citraconic acid, 99+%
Citraconic acid, 99+%
Thermo Scientific Chemicals
Citraconic acid, 99+%
CAS: 498-23-7 | C5H6O4 | 130.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA110431000
Price (MYR)
812.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
812.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCitraconic acid
Name Note99+%
CAS498-23-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: