1-Chloropropane, 99%
1-Chloropropane, 99%
1-Chloropropane, 99%
1-Chloropropane, 99%
Thermo Scientific Chemicals
1-Chloropropane, 99%
CAS: 540-54-5 | C3H7Cl | 78.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA109955000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-chloropropane
CAS540-54-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0