2-Chloro-2-methylpropane, 99%
2-Chloro-2-methylpropane, 99%
2-Chloro-2-methylpropane, 99%
2-Chloro-2-methylpropane, 99%
Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%
2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL | CAS: 507-20-0 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA109505000
Price (MYR)
348.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
348.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Chloro-2-methylpropane
Name Note99%
CAS507-20-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger