5-Chloro-2-methylaniline, 98%
5-Chloro-2-methylaniline, 98%
5-Chloro-2-methylaniline, 98%
5-Chloro-2-methylaniline, 98%
Thermo Scientific Chemicals
5-Chloro-2-methylaniline, 98%
CAS: 95-79-4 | C7H8ClN | 141.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA109380050
Price (MYR)
237.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
237.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Chloro-2-methylaniline
Name Note98%
CAS95-79-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning