1-Chloro-6-hydroxyhexane, 95%
1-Chloro-6-hydroxyhexane, 95%
1-Chloro-6-hydroxyhexane, 95%
1-Chloro-6-hydroxyhexane, 95%
Thermo Scientific Chemicals
1-Chloro-6-hydroxyhexane, 95%
CAS: 2009-83-8 | C6H13ClO | 136.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA109280250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1-chloro-6-hydroxyhexane
CAS2009-83-8
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0