Chlorogenic acid, 98%
Chlorogenic acid, 98%
Chlorogenic acid, 98%
Chlorogenic acid, 98%
Thermo Scientific Chemicals
Chlorogenic acid, 98%
CAS: 327-97-9 | C16H18O9 | 354.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA109240050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChlorogenic acid
Name NotePredominantly trans, From coffee seeds
CAS327-97-9
Melting Point205.0°C to 209.0°C