4-Chlorocinnamic acid, 99%, predominantly trans
4-Chlorocinnamic acid, 99%, predominantly trans
4-Chlorocinnamic acid, 99%, predominantly trans
4-Chlorocinnamic acid, 99%, predominantly trans
Thermo Scientific Chemicals
4-Chlorocinnamic acid, 99%, predominantly trans
CAS: 1615-02-7 | C9H7ClO2 | 182.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA108940250
Price (MYR)
476.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
476.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Chlorocinnamic acid
Name Note99%
CAS1615-02-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning