4-Chlorobenzenesulfonamide, 98+%
4-Chlorobenzenesulfonamide, 98+%
4-Chlorobenzenesulfonamide, 98+%
4-Chlorobenzenesulfonamide, 98+%
Thermo Scientific Chemicals
4-Chlorobenzenesulfonamide, 98+%
CAS: 98-64-6 | C6H6ClNO2S | 191.63 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA108631000100 gPlastic bottle
Catalog number FSA108631000
Price (MYR)
447.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
447.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Chlorobenzenesulfonamide
Name Note98+%
CAS98-64-6
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning