Chloroacetic acid, 99%, flakes
Chloroacetic acid, 99%, flakes
Chloroacetic acid, 99%, flakes
Chloroacetic acid, 99%, flakes
Thermo Scientific Chemicals
Chloroacetic acid, 99%, flakes
CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
4 kg
Packaging:
Plastic bottle
Catalog number FSA108512500
Price (MYR)
234.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
234.00
BOT
Add to cart
Specifications
Chemical Name or MaterialChloroacetic acid
Name Note99%
CAS79-11-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.