N-Carbethoxyphthalimide, 99+%
N-Carbethoxyphthalimide, 99+%
N-Carbethoxyphthalimide, 99+%
N-Carbethoxyphthalimide, 99+%
Thermo Scientific Chemicals
N-Carbethoxyphthalimide, 99+%
CAS: 22509-74-6 | C11H9NO4 | 219.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA108310100
Price (MYR)
271.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
271.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Carbethoxyphthalimide
CAS22509-74-6
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0