S-Butyrylthiocholine iodide, 98%
S-Butyrylthiocholine iodide, 98%
S-Butyrylthiocholine iodide, 98%
S-Butyrylthiocholine iodide, 98%
Thermo Scientific Chemicals
S-Butyrylthiocholine iodide, 98%
CAS: 1866-16-6 | C9H20INOS | 317.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA108150250
Price (MYR)
1,384.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,384.00
BOT
Add to cart
Specifications
Chemical Name or MaterialS-Butyrylthiocholine iodide
CAS1866-16-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.