Butyraldehyde, 99%
Butyraldehyde, 99%
Butyraldehyde, 99%
Thermo Scientific Chemicals
Butyraldehyde, 99%
CAS: 123-72-8 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA108091000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialButyraldehyde
CAS123-72-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.