4-tert-Butylphenol, 97%
4-tert-Butylphenol, 97%
4-tert-Butylphenol, 97%
4-tert-Butylphenol, 97%
Thermo Scientific Chemicals
4-tert-Butylphenol, 97%
CAS: 98-54-4 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
Catalog number FSA108000050
Price (MYR)
172.00
BOT
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
172.00
BOT
Add to cart
Specifications
Chemical Name or Material4-tert-Butylphenol
Name Note97%
CAS98-54-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger