tert-Butylbenzene, 99%
tert-Butylbenzene, 99%
tert-Butylbenzene, 99%
tert-Butylbenzene, 99%
Thermo Scientific Chemicals
tert-Butylbenzene, 99%
CAS: 98-06-6 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number FSA107875000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtert-Butylbenzene
CAS98-06-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.