n-Butylamine, 99+%
n-Butylamine, 99+%
n-Butylamine, 99+%
Thermo Scientific Chemicals
n-Butylamine, 99+%
CAS: 109-73-9 | C4H11N | 73.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA107802500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Butylamine
CAS109-73-9
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0