2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
Thermo Scientific Chemicals
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
CAS: 513-85-9 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA107640010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 3-Butanediol
CAS513-85-9
Melting Point19.0°C
ColorColorless to Yellow
Density1.0100g/mL