alpha-Bromo-p-xylene, 98%
alpha-Bromo-p-xylene, 98%
alpha-Bromo-p-xylene, 98%
alpha-Bromo-p-xylene, 98%
Thermo Scientific Chemicals
alpha-Bromo-p-xylene, 98%
CAS: 104-81-4 | C8H9Br | 185.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA107570250
Price (MYR)
494.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
494.00
BOT
Add to cart
Specifications
Chemical Name or Materialα-Bromo-p-xylene
CAS104-81-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0