3-Bromopyridine, 99%
3-Bromopyridine, 99%
3-Bromopyridine, 99%
3-Bromopyridine, 99%
Thermo Scientific Chemicals
3-Bromopyridine, 99%
CAS: 626-55-1 | C5H4BrN | 158.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA107411000
Price (MYR)
1,285.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,285.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Bromopyridine
Name Note99%
CAS626-55-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger