2-Bromopyridine, 99%
2-Bromopyridine, 99%
2-Bromopyridine, 99%
2-Bromopyridine, 99%
Thermo Scientific Chemicals
2-Bromopyridine, 99%
CAS: 109-04-6 | C5H4BrN | 158 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA107401000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromopyridine
CAS109-04-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0