2-Bromopropane, 99%
2-Bromopropane, 99%
2-Bromopropane, 99%
Thermo Scientific Chemicals
2-Bromopropane, 99%
CAS: 75-26-3 | C3H7Br | 122.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA107322500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromopropane
CAS75-26-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0