4-Bromophenylboronic acid, 98%
4-Bromophenylboronic acid, 98%
4-Bromophenylboronic acid, 98%
4-Bromophenylboronic acid, 98%
Thermo Scientific Chemicals
4-Bromophenylboronic acid, 98%
CAS: 5467-74-3 | C6H6BBrO2 | 200.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA107280250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromophenylboronic acid
CAS5467-74-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0