6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
Thermo Scientific Chemicals
6-Bromo-2-naphthol, 97%
CAS: 15231-91-1 | C10H7BrO | 223.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA107170250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material6-Bromo-2-naphthol
CAS15231-91-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0