(2-Bromoethyl)benzene, 98%
(2-Bromoethyl)benzene, 98%
(2-Bromoethyl)benzene, 98%
(2-Bromoethyl)benzene, 98%
Thermo Scientific Chemicals
(2-Bromoethyl)benzene, 98%
CAS: 103-63-9 | C8H9Br | 185.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA106945000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(2-Bromoethyl)benzene
Name Note98%
CAS103-63-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning