1-Bromo-3-chloropropane, 99%
1-Bromo-3-chloropropane, 99%
1-Bromo-3-chloropropane, 99%
1-Bromo-3-chloropropane, 99%
Thermo Scientific Chemicals
1-Bromo-3-chloropropane, 99%
CAS: 109-70-6 | C3H6BrCl | 157.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA106862500
Price (MYR)
306.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
306.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromo-3-chloropropane
CAS109-70-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0