1-Bromobutane, 99%
1-Bromobutane, 99%
1-Bromobutane, 99%
Thermo Scientific Chemicals
1-Bromobutane, 99%
CAS: 109-65-9 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA106772500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromobutane
CAS109-65-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0