4-Bromobenzoyl chloride, 98%
4-Bromobenzoyl chloride, 98%
4-Bromobenzoyl chloride, 98%
4-Bromobenzoyl chloride, 98%
Thermo Scientific Chemicals
4-Bromobenzoyl chloride, 98%
CAS: 586-75-4 | C7H4BrClO | 219.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA106760250
Price (MYR)
635.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
635.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Bromobenzoyl chloride
CAS586-75-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0