4-Bromobenzonitrile, 98%
4-Bromobenzonitrile, 98%
4-Bromobenzonitrile, 98%
4-Bromobenzonitrile, 98%
Thermo Scientific Chemicals
4-Bromobenzonitrile, 98%
CAS: 623-00-7 | C7H4BrN | 182.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass Bottle
Catalog number FSA106710100
Price (MYR)
319.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
319.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Bromobenzonitrile
Name Note98%
CAS623-00-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning