Bromobenzene, 99%, pure
Bromobenzene, 99%, pure
Bromobenzene, 99%, pure
Bromobenzene, 99%, pure
Thermo Scientific Chemicals
Bromobenzene, 99%, pure
CAS: 108-86-1 | C6H5Br | 157.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA106682500
Price (MYR)
302.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
302.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBromobenzene
CAS108-86-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.