2-Bromoaniline, 98%
2-Bromoaniline, 98%
2-Bromoaniline, 98%
Thermo Scientific Chemicals
2-Bromoaniline, 98%
CAS: 615-36-1 | C6H6BrN | 172.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA106610250
Price (MYR)
436.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
436.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Bromoaniline
CAS615-36-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0